Assay Details
| Gene Symbol: Pcyt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024229.2 | NP_077191.2 | 12 - 13 | 1060 | 75 | |
| GenBank mRNA | AK140458.1 | - | 12 - 13 | 1100 | 75 | |
| AK166319.1 | - | 11 - 12 | 1035 | 75 | ||
| BC003473.1 | - | 12 - 13 | 1066 | 2650077 | 75 | |
| BC008276.1 | - | 12 - 13 | 1060 | 3707732 | 75 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pcyt2
Genomic Map