Assay Details
| Gene Symbol: Tmem79 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024246.5 | NP_077208.1 | 3 - 4 | 1058 | 59 | |
| GenBank mRNA | AK009760.1 | - | 3 - 4 | 1059 | 59 | |
| AK010144.1 | - | 3 - 4 | 1148 | 59 | ||
| BC003309.1 | - | 3 - 4 | 1113 | 3154669 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313147 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313165 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313146 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313144 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313167 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313145 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313166 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148288 | BC003309.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148288 | NM_024246.5 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148288 | AK009760.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148288 | AK010144.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2148288 | AK160793.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 579077 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 209287 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 179390 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P102949 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148288 | BC003309.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148288 | NM_024246.5 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148288 | AK009760.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148288 | AK010144.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2148288 | AK160793.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313150 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313139 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313147 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313138 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313145 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313170 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313142 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313159 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313164 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313165 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313163 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313175 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313154 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313181 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313149 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313180 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313166 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313188 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313184 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313136 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313146 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313190 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313168 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313143 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313177 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313178 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313152 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313160 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313148 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313173 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313153 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313141 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313157 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313158 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313185 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P22313171 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_67_P04980590 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P102949 | - | - |
| Whole Mouse Genome G4122A | A_51_P166873 | BC003309.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P166873 | NM_024246.5 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P166873 | AK009760.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P166873 | AK010144.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P119933 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P102949 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02292924 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03011088 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03011109 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03011107 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03011098 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03011101 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P03075779 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02938391 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02322420 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P02292926 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313145 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313147 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313166 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313165 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313167 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313146 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P22313144 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313208 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313135 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313190 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313180 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313217 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313111 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313131 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313203 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313116 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313185 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313149 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313141 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313169 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313177 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313216 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313153 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313214 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313198 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313095 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313109 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313184 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313157 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313142 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313158 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313150 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313179 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313224 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313145 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313092 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313114 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313118 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313210 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313082 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313193 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313148 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313164 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313154 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313127 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313124 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313160 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313201 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313133 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313212 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313178 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313175 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313166 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313188 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313146 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313163 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313199 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313081 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313204 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313220 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313173 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313110 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313098 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313128 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313191 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313143 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313123 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313126 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313195 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313206 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313168 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313152 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P22313159 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02322420 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313141 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313158 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03011113 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03011087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313131 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03107227 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313148 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03011101 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02685626 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313139 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313133 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02685629 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313147 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02685623 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03011086 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313152 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313135 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313130 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313143 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P03011088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313150 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313124 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02685631 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P02685622 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313154 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313145 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P22313142 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10499353 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499349 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499350 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499351 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10499352 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1430483_PM_a_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10499348 | - | - |
| Mouse Genome 430 2.0 Array | 1430483_PM_a_at | - | - |
| Mouse Genome 430 2.0 Array | 1430483_a_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71913_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 71913_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0300017454.mm.1 | NM_024246.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017454.mm.1 | NM_024246.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017454.mm.1 | TC0300002306.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0300017451.mm.1 | NM_024246.5.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017451.mm.1 | NM_024246.5 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0300017451.mm.1 | TC0300002306.mm.1 | MTA TC |
| Murine Genome U74v2 Set | 167778_i_at | - | - |
| Murine Genome U74v2 Set | 170263_f_at | - | - |
| Murine Genome U74v2 Set | 170263_PM_f_at | - | - |
| Murine Genome U74v2 Set | 167778_PM_i_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430483_a_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1430483_PM_a_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 915881 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6940288 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006940288 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_1231048 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_1231048 | - | - |
| mouse-6 v1.1 (Illumina) | scl21978.7.1_53-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6940288 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl21978.7.1_53-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6940288 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_1231048 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_1231048 | - | - |

Tmem79
Genomic Map