Assay Details
| Gene Symbol: Aplf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170489.1 | NP_001163960.1 | 3 - 4 | 545 | 257 | |
| NM_024251.4 | NP_077213.2 | 3 - 4 | 415 | 257 | ||
| GenBank mRNA | AK008513.1 | - | 3 - 4 | 546 | 257 | |
| AK034191.1 | - | 3 - 4 | 395 | 257 | ||
| BC002179.1 | - | 3 - 4 | 193 | 3487226 | 115 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the BC002179.1 transcripts.
Set Membership: |
|

Aplf
Genomic Map