Assay Details
| Gene Symbol: Scd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024450.2 | NP_077770.1 | 2 - 3 | 543 | 68 | |
| GenBank mRNA | AF272037.1 | - | 2 - 3 | 532 | 68 | |
| AK078675.1 | - | 2 - 3 | 566 | 68 | ||
| AK078713.1 | - | 2 - 3 | 565 | 68 | ||
| AK136277.1 | - | 2 - 3 | 560 | 68 | ||
| BC118033.1 | - | 2 - 3 | 441 | 40106893 | 68 | |
| BC118045.1 | - | 2 - 3 | 441 | 40106896 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scd3
Genomic Map