Assay Details
| Gene Symbol: Cyc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025567.2 | NP_079843.1 | 2 - 3 | 370 | 61 | |
| GenBank mRNA | AK002815.1 | - | 2 - 3 | 348 | 61 | |
| AK011288.1 | - | 2 - 3 | 371 | 61 | ||
| AK151299.1 | - | 2 - 3 | 361 | 61 | ||
| AK170272.1 | - | 1 - 2 | 970 | 61 | ||
| AK184866.1 | - | 1 - 2 | 124 | 61 | ||
| AK188690.1 | - | 1 - 2 | 126 | 61 | ||
| AK192516.1 | - | 2 - 3 | 226 | 61 | ||
| AK195664.1 | - | 1 - 2 | 124 | 61 | ||
| AK215816.1 | - | 1 - 2 | 124 | 61 | ||
| BC005620.1 | - | 2 - 3 | 351 | 3711323 | 61 | |
| BC082790.1 | - | 2 - 3 | 308 | 61 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyc1
Genomic Map