Assay Details
| Gene Symbol: Cyc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025567.2 | NP_079843.1 | 3 - 4 | 498 | 88 | |
| GenBank mRNA | AK002815.1 | - | 3 - 4 | 476 | 88 | |
| AK011288.1 | - | 3 - 4 | 499 | 88 | ||
| AK151299.1 | - | 3 - 4 | 489 | 88 | ||
| AK170272.1 | - | 2 - 3 | 1098 | 88 | ||
| AK192516.1 | - | 3 - 4 | 355 | 88 | ||
| AK215816.1 | - | 2 - 3 | 251 | 88 | ||
| BC005620.1 | - | 3 - 4 | 479 | 3711323 | 88 | |
| BC082790.1 | - | 3 - 4 | 436 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyc1
Genomic Map