Assay Details
| Gene Symbol: Cyc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025567.2 | NP_079843.1 | 4 - 5 | 655 | 66 | |
| GenBank mRNA | AK002815.1 | - | 4 - 5 | 633 | 66 | |
| AK011288.1 | - | 4 - 5 | 656 | 66 | ||
| AK151299.1 | - | 4 - 5 | 646 | 66 | ||
| AK170272.1 | - | 3 - 4 | 1255 | 66 | ||
| BC005620.1 | - | 4 - 5 | 636 | 3711323 | 66 | |
| BC082790.1 | - | 4 - 5 | 593 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cyc1
Genomic Map