Assay Details
| Gene Symbol: Fbxo25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025785.2 | NP_080061.1 | 1 - 2 | 61 | 61 | |
| GenBank mRNA | AK008027.1 | - | 1 - 2 | 65 | 61 | |
| AK013645.1 | - | 1 - 2 | 62 | 61 | ||
| AK018626.1 | - | 1 - 2 | 62 | 61 | ||
| AK134722.1 | - | 1 - 2 | 65 | 61 | ||
| BC014749.1 | - | 1 - 2 | 47 | 4240953 | 61 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK008027.1,AK018626.1,AK134722.1,BC014749.1,NM_025785.2 transcripts.
Set Membership: |
|

Fbxo25
Genomic Map