Assay Details
Gene Symbol: Psmd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_025894.2 | NP_080170.1 | 5 - 6 | 559 | 80 | |
GenBank mRNA | AK005114.1 | - | 5 - 6 | 594 | 80 | |
AK007619.1 | - | 5 - 6 | 604 | 80 | ||
AK007680.1 | - | 5 - 6 | 533 | 80 | ||
AK008646.1 | - | 5 - 6 | 560 | 80 | ||
AK009969.1 | - | 5 - 6 | 562 | 80 | ||
AK011806.1 | - | 5 - 6 | 583 | 80 | ||
AK017843.1 | - | 5 - 6 | 576 | 80 | ||
AK150910.1 | - | 5 - 6 | 583 | 80 | ||
AK151242.1 | - | 5 - 6 | 560 | 80 | ||
AK152041.1 | - | 5 - 6 | 583 | 80 | ||
AK152240.1 | - | 5 - 6 | 591 | 80 | ||
AK160283.1 | - | 5 - 6 | 581 | 80 | ||
AK162779.1 | - | 560 | 80 | |||
AK167236.1 | - | 5 - 6 | 567 | 80 | ||
AK183870.1 | - | 2 - 3 | 126 | 80 | ||
AK186271.1 | - | 2 - 3 | 126 | 80 | ||
BC004694.1 | - | 5 - 6 | 577 | 3498952 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
