Assay Details
Gene Symbol: Ergic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026170.3 | NP_080446.1 | 3 - 4 | 256 | 64 | |
GenBank mRNA | AB093571.1 | - | 3 - 4 | 155 | 64 | |
AK003239.1 | - | 3 - 4 | 243 | 64 | ||
AK004629.1 | - | 3 - 4 | 210 | 64 | ||
AK084385.1 | - | 3 - 4 | 203 | 64 | ||
AK088157.1 | - | 3 - 4 | 211 | 64 | ||
AK151239.1 | - | 3 - 4 | 210 | 64 | ||
AK173123.1 | - | 3 - 4 | 245 | 64 | ||
BC005516.1 | - | 3 - 4 | 256 | 3494155 | 64 | |
BC083144.1 | - | 3 - 4 | 176 | 30432732 | 64 | |
CT010230.1 | - | 3 - 4 | 155 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
