Assay Details
Gene Symbol: Ergic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026170.3 | NP_080446.1 | 5 - 6 | 482 | 68 | |
GenBank mRNA | AB093571.1 | - | 5 - 6 | 381 | 68 | |
AK004629.1 | - | 5 - 6 | 436 | 68 | ||
AK040727.1 | - | 2 - 3 | 634 | 68 | ||
AK084385.1 | - | 5 - 6 | 429 | 68 | ||
AK088157.1 | - | 5 - 6 | 437 | 68 | ||
AK151239.1 | - | 5 - 6 | 436 | 68 | ||
AK173123.1 | - | 5 - 6 | 471 | 68 | ||
BC005516.1 | - | 5 - 6 | 482 | 3494155 | 68 | |
BC083144.1 | - | 5 - 6 | 402 | 30432732 | 68 | |
CT010230.1 | - | 5 - 6 | 381 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
