Assay Details
Gene Symbol: Ergic1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026170.3 | NP_080446.1 | 7 - 8 | 645 | 121 | |
GenBank mRNA | AB093571.1 | - | 7 - 8 | 544 | 121 | |
AK003239.1 | - | 7 - 8 | 631 | 121 | ||
AK004629.1 | - | 7 - 8 | 599 | 121 | ||
AK040727.1 | - | 4 - 5 | 797 | 121 | ||
AK084385.1 | - | 7 - 8 | 592 | 121 | ||
AK088157.1 | - | 7 - 8 | 600 | 121 | ||
AK151239.1 | - | 7 - 8 | 599 | 121 | ||
AK173123.1 | - | 7 - 8 | 634 | 121 | ||
AK196223.1 | - | 2 - 3 | 114 | 121 | ||
AK196818.1 | - | 2 - 3 | 114 | 121 | ||
AK198608.1 | - | 2 - 3 | 121 | |||
AK204168.1 | - | 2 - 3 | 114 | 121 | ||
AK204170.1 | - | 2 - 3 | 114 | 121 | ||
BC005516.1 | - | 7 - 8 | 645 | 3494155 | 121 | |
BC083144.1 | - | 7 - 8 | 565 | 30432732 | 121 | |
CT010230.1 | - | 7 - 8 | 544 | 121 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
