Assay Details
| Gene Symbol: Mmd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026178.2 | NP_080454.1 | 5 - 6 | 677 | 91 | |
| GenBank mRNA | AK004819.1 | - | 5 - 6 | 631 | 91 | |
| AK011418.1 | - | 5 - 6 | 642 | 91 | ||
| AK033889.1 | - | 5 - 6 | 697 | 91 | ||
| AK149704.1 | - | 5 - 6 | 627 | 91 | ||
| AK158577.1 | - | 5 - 6 | 635 | 91 | ||
| AK171637.1 | - | 5 - 6 | 631 | 91 | ||
| AY424300.1 | - | 5 - 6 | 517 | 91 | ||
| BC021914.1 | - | 5 - 6 | 677 | 4506710 | 91 | |
| BC034117.1 | - | 6 - 7 | 1029 | 91 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Mmd
Genomic Map