Assay Details
| Gene Symbol: Abhd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026179.2 | NP_080455.1 | 1 - 2 | 224 | 105 | |
| GenBank mRNA | AK004873.1 | - | 1 - 2 | 110 | 105 | |
| AK050377.1 | - | 1 - 2 | 109 | 105 | ||
| BC037063.1 | - | 1 - 2 | 106 | 4985992 | 105 | |
| CN840045.1 | - | 1 - 2 | 235 | 105 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK004873.1,AK050377.1,BC037063.1,NM_026179.2 transcripts.
Set Membership: |
|

Abhd5
Genomic Map