Assay Details
| Gene Symbol: Abhd5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026179.2 | NP_080455.1 | 5 - 6 | 950 | 87 | |
| GenBank mRNA | AK004873.1 | - | 5 - 6 | 836 | 87 | |
| AK007421.1 | - | 5 - 6 | 87 | |||
| AK019488.2 | - | 3 - 4 | 376 | 87 | ||
| AK028453.1 | - | 5 - 6 | 836 | 87 | ||
| AK050377.1 | - | 5 - 6 | 835 | 87 | ||
| AK152717.1 | - | 6 - 7 | 884 | 87 | ||
| BC006683.1 | - | 2 - 3 | 176 | 87 | ||
| BC037063.1 | - | 5 - 6 | 832 | 4985992 | 87 | |
More Information
Updated Mapping Information
Set Membership: |
|

Abhd5
Genomic Map