Assay Details
| Gene Symbol: Glipr1l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026223.2 | NP_080499.1 | 4 - 5 | 693 | 83 | |
| GenBank mRNA | AK014846.1 | - | 4 - 5 | 694 | 83 | |
| AK015834.1 | - | 4 - 5 | 677 | 83 | ||
| AK161292.1 | - | 4 - 5 | 694 | 83 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Glipr1l2
Genomic Map