Assay Details
| Gene Symbol: Slc25a26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026255.5 | NP_080531.2 | 4 - 5 | 522 | 121 | |
| GenBank mRNA | AK015299.1 | - | 4 - 5 | 504 | 121 | |
| AK015954.1 | - | 4 - 5 | 541 | 121 | ||
| BC019156.1 | - | 5 - 6 | 492 | 5066084 | 121 | |
| BC019170.1 | - | 4 - 5 | 523 | 5003084 | 121 | |
| BC037142.1 | - | 4 - 5 | 499 | 5026631 | 121 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a26
Genomic Map