Assay Details
| Gene Symbol: Slc25a26 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026255.5 | NP_080531.2 | 5 - 6 | 565 | 80 | |
| GenBank mRNA | AK015299.1 | - | 5 - 6 | 547 | 80 | |
| AK015954.1 | - | 5 - 6 | 584 | 80 | ||
| BC019156.1 | - | 6 - 7 | 535 | 5066084 | 80 | |
| BC019170.1 | - | 5 - 6 | 566 | 5003084 | 80 | |
| BC037142.1 | - | 5 - 6 | 542 | 5026631 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a26
Genomic Map