Assay Details
| Gene Symbol: Tctn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026260.2 | NP_080536.2 | 8 - 9 | 1004 | 66 | |
| GenBank mRNA | AK015861.1 | - | 8 - 9 | 992 | 66 | |
| AK016982.1 | - | 8 - 9 | 990 | 66 | ||
| AK030156.1 | - | 8 - 9 | 990 | 66 | ||
| BC027335.1 | - | 8 - 9 | 1004 | 3710046 | 66 | |
| BC080805.1 | - | 8 - 9 | 979 | 5352578 | 66 | |
| DQ278871.1 | - | 8 - 9 | 1021 | 66 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tctn3
Genomic Map