Assay Details
| Gene Symbol: Rpp21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026308.2 | NP_080584.1 | 1 - 2 | 69 | 105 | |
| GenBank mRNA | AJ504717.1 | - | 1 - 2 | 79 | 105 | |
| AK002780.1 | - | 1 - 2 | 70 | 105 | ||
| AK003455.1 | - | 1 - 2 | 67 | 105 | ||
| BC028491.1 | - | 1 - 2 | 79 | 1497187 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rpp21
Genomic Map