Assay Details
| Gene Symbol: Tmem179b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026325.3 | NP_080601.2 | 1 - 2 | 137 | 92 | |
| GenBank mRNA | AK007906.1 | - | 1 - 2 | 113 | 92 | |
| AK010996.1 | - | 1 - 2 | 118 | 92 | ||
| BC116961.1 | - | 1 - 2 | 137 | 40126280 | 92 | |
| BC116963.1 | - | 1 - 2 | 137 | 40126282 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31943526 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31943527 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P228031 | BC061068.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P228031 | BC087914.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P228031 | BC116961.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P228031 | BC116963.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P228031 | BC144813.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P228031 | BC061068.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P228031 | BC087914.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P228031 | BC116961.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P228031 | BC116963.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_52_P228031 | BC144813.1 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 850449 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P228031 | BC061068.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P228031 | BC087914.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P228031 | BC116961.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P228031 | BC116963.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P228031 | BC144813.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P228031 | BC061068.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P228031 | BC087914.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P228031 | BC116961.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P228031 | BC116963.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_52_P228031 | BC144813.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943525 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943526 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943515 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943520 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943522 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943523 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943519 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943513 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943518 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943521 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P31943516 | - | - |
| Whole Mouse Genome G4122A | A_51_P416625 | BC061068.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P416625 | BC087914.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P416625 | BC116961.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P416625 | BC116963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P416625 | BC144813.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P228031 | BC061068.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P228031 | BC087914.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P228031 | BC116961.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P228031 | BC116963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P228031 | BC144813.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P228031 | BC061068.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P228031 | BC087914.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P228031 | BC116961.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P228031 | BC116963.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P228031 | BC144813.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P137028 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P114901 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14384254 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31943526 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P31943527 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943436 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943455 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943450 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943453 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943501 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943516 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943526 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943443 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943438 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943515 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943505 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943433 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943522 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943519 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943503 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943520 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943440 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943525 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943454 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943521 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943437 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943447 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943513 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943504 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P31943518 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31943520 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31943511 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31943518 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31943519 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31943522 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31943515 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14384249 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31943525 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31943526 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P31943513 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10465787 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465784 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465789 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465785 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465788 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465791 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10465786 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1455874_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449842_PM_at | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1436963_PM_x_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10465790 | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10465783 | - | - |
| Mouse Genome 430 2.0 Array | 1436963_PM_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1449842_at | - | - |
| Mouse Genome 430 2.0 Array | 1455874_at | - | - |
| Mouse Genome 430 2.0 Array | 1436963_x_at | - | - |
| Mouse Genome 430 2.0 Array | 1455874_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449842_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67706_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 67706_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1900009982.mm.1 | NM_026325.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009982.mm.1 | NM_026325.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1900009982.mm.1 | TC1900001769.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1455874_PM_at | - | - |
| Mouse Expression Set 430 | 1455874_at | - | - |
| Mouse Expression Set 430 | 1436963_PM_x_at | - | - |
| Mouse Expression Set 430 | 1449842_PM_at | - | - |
| Mouse Expression Set 430 | 1436963_x_at | - | - |
| Mouse Expression Set 430 | 1449842_at | - | - |
| Murine Genome U74v2 Set | 160497_at | - | - |
| Murine Genome U74v2 Set | 160497_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455874_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436963_PM_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1436963_x_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449842_at | - | - |
| Mouse Genome 430A 2.0 Array | 1455874_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449842_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455874_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436963_PM_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1436963_x_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1455874_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449842_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449842_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 930156 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Illumina mouse-6 v1.0 expression beadchip | 6860064 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0006860064 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2656071 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2656071 | - | - |
| mouse-6 v1.1 (Illumina) | scl067706.2_5-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 6860064 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl067706.2_5-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 6860064 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2656071 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2656071 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl067706.2_5-S | - | - |

Tmem179b
Genomic Map