Assay Details
| Gene Symbol: Ilf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026374.3 | NP_080650.1 | 11 - 12 | 848 | 58 | |
| GenBank mRNA | AK013858.1 | - | 11 - 12 | 894 | 58 | |
| AK078003.1 | - | 11 - 12 | 835 | 58 | ||
| AK135539.1 | - | 11 - 12 | 835 | 58 | ||
| AK157129.1 | - | 11 - 12 | 903 | 58 | ||
| BC004033.1 | - | 3 - 4 | 215 | 58 | ||
| BC024718.1 | - | 11 - 12 | 848 | 5012365 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ilf2
Genomic Map