Assay Details
| Gene Symbol: Rnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170953.1 | NP_001164424.1 | 1 - 2 | 71 | 73 | |
| NM_026440.4 | NP_080716.1 | 1 - 2 | 71 | 73 | ||
| GenBank mRNA | AK011300.1 | - | 1 - 2 | 55 | 73 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK011300.1,NM_001170953.1,NM_026440.4 transcripts.
Set Membership: |
|

Rnmt
Genomic Map