Assay Details
| Gene Symbol: Rnmt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170953.1 | NP_001164424.1 | 8 - 9 | 1274 | 93 | |
| NM_026440.4 | NP_080716.1 | 9 - 10 | 1439 | 93 | ||
| GenBank mRNA | AK011300.1 | - | 9 - 10 | 1423 | 93 | |
| AK015403.1 | - | 8 - 9 | 1231 | 93 | ||
| AK082082.1 | - | 1 - 2 | 224 | 93 | ||
| AK082331.1 | - | 9 - 10 | 1406 | 93 | ||
| AK129130.1 | - | 8 - 9 | 1376 | 93 | ||
| BC021794.1 | - | 9 - 10 | 1385 | 4208274 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rnmt
Genomic Map