Assay Details
| Gene Symbol: Chid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142681.1 | NP_001136153.1 | 8 - 9 | 829 | 118 | |
| GenBank mRNA | AK014071.1 | - | 8 - 9 | 778 | 118 | |
| AK036717.1 | - | 8 - 9 | 774 | 118 | ||
| AK147377.1 | - | 6 - 7 | 5627 | 118 | ||
| AK160156.1 | - | 8 - 9 | 789 | 118 | ||
| AK160200.1 | - | 8 - 9 | 790 | 118 | ||
| AK193697.1 | - | 3 - 4 | 215 | 118 | ||
| BC006876.1 | - | 8 - 9 | 778 | 2647540 | 118 | |
| BC061063.1 | - | 8 - 9 | 789 | 6591338 | 118 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chid1
Genomic Map