Assay Details
| Gene Symbol: Chid1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001142681.1 | NP_001136153.1 | 11 - 12 | 1164 | 80 | |
| NM_026522.5 | NP_080798.2 | 9 - 10 | 906 | 80 | ||
| GenBank mRNA | AK014071.1 | - | 11 - 12 | 1113 | 80 | |
| AK036717.1 | - | 11 - 12 | 1109 | 80 | ||
| AK050655.1 | - | 9 - 10 | 854 | 80 | ||
| AK147377.1 | - | 9 - 10 | 5962 | 80 | ||
| AK160156.1 | - | 11 - 12 | 1124 | 80 | ||
| AK160200.1 | - | 11 - 12 | 1125 | 80 | ||
| BC006876.1 | - | 11 - 12 | 1113 | 2647540 | 80 | |
| BC061063.1 | - | 11 - 12 | 1124 | 6591338 | 80 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chid1
Genomic Map