Assay Details
| Gene Symbol: Slc25a39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026542.3 | NP_080818.1 | 10 - 11 | 1024 | 100 | |
| GenBank mRNA | AK007934.1 | - | 10 - 11 | 978 | 100 | |
| AK013834.1 | - | 6 - 7 | 668 | 100 | ||
| AK078417.1 | - | 10 - 11 | 1016 | 100 | ||
| AK160922.1 | - | 10 - 11 | 970 | 100 | ||
| AK161289.1 | - | 10 - 11 | 1016 | 100 | ||
| BC115661.1 | - | 10 - 11 | 924 | 40103267 | 100 | |
| BC115662.1 | - | 10 - 11 | 931 | 40103268 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Slc25a39
Genomic Map