Assay Details
| Gene Symbol: Sult1c2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026935.4 | NP_081211.3 | 1 - 2 | 181 | 66 | |
| GenBank mRNA | AK007384.1 | - | 1 - 2 | 41 | 66 | |
| AK144336.1 | - | 1 - 2 | 121 | 66 | ||
| AY005469.1 | - | 1 - 2 | 181 | 66 | ||
| BC022665.1 | - | 1 - 2 | 174 | 4223678 | 66 | |
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK007384.1,AK144336.1,AY005469.1,BC022665.1,NM_026935.4 transcripts.
Set Membership: |
|

Sult1c2
Genomic Map