Assay Details
| Gene Symbol: Ufsp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027356.2 | NP_081632.1 | 1 | 251 | 59 | |
| GenBank mRNA | AK012343.1 | - | 1 | 252 | 59 | |
| BC016577.1 | - | 1 | 260 | 3584484 | 59 | |
| BC087958.1 | - | 1 | 272 | 6751787 | 59 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ufsp1
Genomic Map