Assay Details
| Gene Symbol: Akr1cl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027582.3 | NP_081858.2 | 5 - 6 | 650 | 114 | |
| GenBank mRNA | AK014937.1 | - | 4 - 5 | 529 | 114 | |
| AK135492.1 | - | 5 - 6 | 653 | 114 | ||
| BC051128.1 | - | 5 - 6 | 650 | 6707386 | 114 | |
More Information
Updated Mapping Information
Set Membership: |
|

Akr1cl
Genomic Map