Assay Details
| Gene Symbol: Actrt1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028514.3 | NP_082790.1 | 1 | 655 | 63 | |
| GenBank mRNA | AK006855.1 | - | 1 | 656 | 63 | |
| BC116431.1 | - | 1 | 630 | 40104427 | 63 | |
| BC116432.1 | - | 1 | 630 | 40104435 | 63 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518094 | BC116432.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518094 | NM_028514.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518094 | AK006855.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518094 | BC116431.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518094 | BC116432.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518094 | NM_028514.3 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518094 | AK006855.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P518094 | BC116431.1 | mouse_transcriptome |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P147915 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518094 | BC116432.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518094 | NM_028514.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518094 | AK006855.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518094 | BC116431.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518094 | BC116432.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518094 | NM_028514.3 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518094 | AK006855.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P518094 | BC116431.1 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32355670 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32355678 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32355683 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32355672 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32355675 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32355680 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32355686 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32355684 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32355673 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32355685 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32355671 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32355681 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32355688 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32355687 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32355677 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P32355676 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P147915 | - | - |
| Whole Mouse Genome G4122A | A_51_P518094 | BC116432.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518094 | NM_028514.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518094 | AK006855.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518094 | BC116431.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518094 | BC116432.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518094 | NM_028514.3 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518094 | AK006855.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P518094 | BC116431.1 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P124067 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P147915 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14635155 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P14635161 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355672 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355696 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355700 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355680 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355681 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355692 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355685 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355684 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355694 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355695 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355693 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355698 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355673 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355678 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355683 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355687 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355689 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355675 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P32355688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14635167 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14635152 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P14635160 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32355688 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32355684 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32355691 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32355683 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32355687 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32355678 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32355681 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32355676 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32355669 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32355680 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32355672 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32355668 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32355689 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32355685 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P32355673 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10599426 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449825_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10599425 | - | - |
| Mouse Genome 430 2.0 Array | 1449825_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449825_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73360_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 73360_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0X00002759.mm.1 | NM_028514.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00002759.mm.1 | NM_028514.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00002759.mm.1 | TC0X00000418.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0X00002758.mm.1 | NM_028514.3.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00002758.mm.1 | NM_028514.3 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0X00002758.mm.1 | TC0X00000418.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449825_PM_at | - | - |
| Mouse Expression Set 430 | 1449825_at | - | - |
| Murine Genome U74v2 Set | 167445_at | - | - |
| Murine Genome U74v2 Set | 167445_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449825_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449825_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449825_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449825_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 778744 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4480347 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4480347 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480347 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2618100 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2618100 | - | - |
| mouse-6 v1.1 (Illumina) | scl54955.1.1_139-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480347 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl54955.1.1_139-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480347 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2618100 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2618100 | - | - |

Actrt1
Genomic Map