Assay Details
| Gene Symbol: Pop7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028753.2 | NP_083029.1 | 1 - 2 | 154 | 124 | |
| GenBank mRNA | AK002782.1 | - | 1 - 2 | 156 | 124 | |
| AK162268.1 | - | 1 - 2 | 159 | 124 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AK002782.1,AK162268.1,NM_028753.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126055 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126061 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126059 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126098 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126096 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126095 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126100 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126060 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126058 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126094 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126062 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126056 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126057 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047698 | AK002782.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047698 | AK158620.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2047698 | NM_028753.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 229531 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P140410 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047698 | AK002782.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047698 | AK158620.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2047698 | NM_028753.2 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P307467 | AK162268.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P307467 | BC010780.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P307467 | AK002782.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P307467 | AK158620.1 | mouse_transcriptome |
| Agilent Genotypic Technology designed "Custom Agilent Mus Musculus 8x15k GE Microarray (AMADID-016270) " | A_51_P307467 | NM_028753.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126056 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126079 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126078 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126045 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126070 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126064 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126075 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126060 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126044 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126063 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126057 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126050 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126071 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126054 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126048 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126074 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126066 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126049 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126052 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P24126055 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P140410 | - | - |
| Whole Mouse Genome G4122A | A_51_P307467 | AK162268.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307467 | BC010780.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307467 | AK002782.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307467 | AK158620.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P307467 | NM_028753.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P113379 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P140410 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04448333 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04448324 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04448337 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04291836 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04448326 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P04448345 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126094 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126059 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126058 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126096 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126098 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126100 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126056 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126095 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126055 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126062 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126060 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126057 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P24126061 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126074 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126078 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126050 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126056 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126091 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126094 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126060 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126089 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126066 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126064 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126046 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126048 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126055 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126103 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126067 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126097 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126100 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126063 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126096 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126088 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126075 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126087 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126057 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126099 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126105 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126093 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126022 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126101 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126049 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126070 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126035 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126037 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126084 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126024 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126052 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P24126044 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04526986 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126060 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126041 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04663136 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126087 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126035 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04448324 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126039 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04692653 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04294715 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126074 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126070 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04448337 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04904360 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126055 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04831288 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126050 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126079 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126076 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04448340 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126036 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126093 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126082 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126048 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04983366 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126066 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126042 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04296014 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126084 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126064 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126092 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04927800 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126043 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04448338 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04448334 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04293356 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126088 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126054 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126037 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P04526990 | - | - |
| Agilent Six_ChIP_01_of_03_v2 | A_68_P24126078 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10534840 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10534841 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449162_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10534839 | - | - |
| Mouse Genome 430 2.0 Array | 1449162_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449162_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74097_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 74097_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0500031400.mm.1 | TC0500003448.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500031400.mm.1 | NM_028753.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031400.mm.1 | NM_028753.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031402.mm.1 | TC0500003448.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500031402.mm.1 | NM_028753.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031402.mm.1 | NM_028753.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031403.mm.1 | TC0500003448.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0500031403.mm.1 | NM_028753.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0500031403.mm.1 | NM_028753.2 | Refseq |
| Mouse Expression Set 430 | 1449162_PM_at | - | - |
| Mouse Expression Set 430 | 1449162_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449162_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449162_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449162_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449162_PM_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 653392 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 380735 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 380735 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0000380735 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2619362 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2619362 | - | - |
| mouse-6 v1.1 (Illumina) | scl25877.2.1_20-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 380735 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl25877.2.1_20-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 380735 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2619362 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2619362 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl25877.2.1_20-S | - | - |

Pop7
Genomic Map