Assay Details
| Gene Symbol: Car15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030558.2 | NP_085035.1 | 7 - 8 | 842 | 58 | |
| GenBank mRNA | AF231122.1 | - | 7 - 8 | 794 | 58 | |
| AK131988.1 | - | 7 - 8 | 870 | 58 | ||
| BC019975.1 | - | 7 - 8 | 842 | 4223351 | 58 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154416 | AF231122.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154416 | BC019975.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_55_P2154416 | NM_030558.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 150690 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154416 | AF231122.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154416 | BC019975.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_55_P2154416 | NM_030558.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576007 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30575994 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576018 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576026 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30575997 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30575988 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30575989 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30575993 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30575995 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576011 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576019 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576002 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576013 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576021 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576023 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30575990 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576009 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576016 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576008 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30575991 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576003 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576001 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30575996 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576014 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30576004 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P30575998 | - | - |
| Whole Mouse Genome G4122A | A_52_P445853 | AF231122.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P445853 | AK131988.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P445853 | BC019975.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_52_P445853 | NM_030558.2 | mouse_transcriptome |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12850406 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12511555 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12860643 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12511566 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12511567 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12511568 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12860641 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12511563 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12676423 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P12860637 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575934 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575980 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575958 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575996 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575952 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576002 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575974 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575995 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575931 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575971 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575955 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576026 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576004 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575959 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575965 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576014 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575946 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575972 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575985 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575936 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575991 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575953 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576021 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575951 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575948 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575977 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576001 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575957 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576016 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575926 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576019 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575967 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575986 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575950 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575988 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576023 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575994 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575956 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575998 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575993 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576008 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575930 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575983 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575981 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575962 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575979 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30575929 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576007 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576011 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P30576013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12860641 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30576013 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30575988 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30575956 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30576007 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30575989 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12511557 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30575955 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13028731 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30575979 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13028734 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30575967 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12850406 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30575966 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30575986 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30576021 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30575969 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30576008 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30576022 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12676419 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30576004 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13028732 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12511571 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30575959 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12511568 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30575957 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30575991 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30575958 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30576026 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30575965 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13028743 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30576015 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30576016 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30575981 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30575996 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12511565 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30576001 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P12860643 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13028729 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30576023 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P30575970 | - | - |
| Agilent Six_ChIP_03_of_03_v2 | A_68_P13028740 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10438226 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438229 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438231 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438227 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438221 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438223 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438225 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438222 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10438230 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1449234_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10438220 | - | - |
| Mouse Genome 430 2.0 Array | 1449234_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1449234_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 80733_PM_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 80733_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR1600016806.mm.1 | NM_030558.2.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600016806.mm.1 | NM_030558.2 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR1600016806.mm.1 | TC1600002180.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1449234_at | - | - |
| Mouse Expression Set 430 | 1449234_PM_at | - | - |
| Murine Genome U74v2 Set | 168374_PM_at | - | - |
| Murine Genome U74v2 Set | 168374_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449234_PM_at | - | - |
| Mouse Genome 430A 2.0 Array | 1449234_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449234_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1449234_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| ABI Mouse Genome Survey Microarray | 562473 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 4480497 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 4480497 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0004480497 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2685543 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2685543 | - | - |
| mouse-6 v1.1 (Illumina) | scl080733.2_18-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 4480497 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl080733.2_18-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 4480497 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2871660 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2871660 | - | - |

Car15
Genomic Map