Assay Details
| Gene Symbol: Sacm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030692.4 | NP_109617.1 | 2 - 3 | 136 | 104 | |
| GenBank mRNA | AJ245720.1 | - | 2 - 3 | 266 | 104 | |
| AJ297526.1 | - | 2 - 3 | 137 | 104 | ||
| AK089174.1 | - | 2 - 3 | 220 | 104 | ||
| AK122390.1 | - | 2 - 3 | 201 | 104 | ||
| AK154759.1 | - | 2 - 3 | 237 | 104 | ||
| AK161111.1 | - | 2 - 3 | 170 | 104 | ||
| AK171080.1 | - | 2 - 3 | 220 | 104 | ||
| BC117752.1 | - | 2 - 3 | 137 | 40090556 | 104 | |
| BC117753.1 | - | 2 - 3 | 137 | 40090558 | 104 | |
| EF437383.1 | - | 2 - 3 | 185 | 104 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sacm1l
Genomic Map