Assay Details
| Gene Symbol: Sacm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030692.4 | NP_109617.1 | 7 - 8 | 580 | 100 | |
| GenBank mRNA | AJ245720.1 | - | 7 - 8 | 710 | 100 | |
| AJ297526.1 | - | 7 - 8 | 581 | 100 | ||
| AK089174.1 | - | 7 - 8 | 664 | 100 | ||
| AK122390.1 | - | 7 - 8 | 645 | 100 | ||
| AK154759.1 | - | 7 - 8 | 680 | 100 | ||
| AK161111.1 | - | 7 - 8 | 614 | 100 | ||
| AK171080.1 | - | 7 - 8 | 664 | 100 | ||
| BC117752.1 | - | 7 - 8 | 581 | 40090556 | 100 | |
| BC117753.1 | - | 7 - 8 | 581 | 40090558 | 100 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sacm1l
Genomic Map