Assay Details
| Gene Symbol: Sacm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030692.4 | NP_109617.1 | 10 - 11 | 861 | 67 | |
| GenBank mRNA | AJ245720.1 | - | 10 - 11 | 991 | 67 | |
| AJ297526.1 | - | 10 - 11 | 862 | 67 | ||
| AK089174.1 | - | 10 - 11 | 945 | 67 | ||
| AK122390.1 | - | 10 - 11 | 926 | 67 | ||
| AK154759.1 | - | 10 - 11 | 961 | 67 | ||
| AK161111.1 | - | 10 - 11 | 895 | 67 | ||
| AK171080.1 | - | 10 - 11 | 945 | 67 | ||
| BC005489.1 | - | 1 - 2 | 70 | 67 | ||
| BC117752.1 | - | 10 - 11 | 862 | 40090556 | 67 | |
| BC117753.1 | - | 10 - 11 | 862 | 40090558 | 67 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sacm1l
Genomic Map