Assay Details
| Gene Symbol: Sacm1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030692.4 | NP_109617.1 | 18 - 19 | 1577 | 83 | |
| GenBank mRNA | AJ245720.1 | - | 18 - 19 | 1707 | 83 | |
| AJ297526.1 | - | 18 - 19 | 1578 | 83 | ||
| AK089174.1 | - | 18 - 19 | 1661 | 83 | ||
| AK122390.1 | - | 18 - 19 | 1642 | 83 | ||
| AK154759.1 | - | 18 - 19 | 1677 | 83 | ||
| AK161111.1 | - | 18 - 19 | 1611 | 83 | ||
| AK171080.1 | - | 18 - 19 | 1661 | 83 | ||
| BC005489.1 | - | 9 - 10 | 786 | 83 | ||
| BC117752.1 | - | 18 - 19 | 1578 | 40090556 | 83 | |
| BC117753.1 | - | 18 - 19 | 1578 | 40090558 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Sacm1l
Genomic Map