Assay Details
| Gene Symbol: Maged2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199246.1 | NP_001186175.1 | 7 - 8 | 1195 | 64 | |
| NM_030700.2 | NP_109625.1 | 7 - 8 | 1218 | 64 | ||
| GenBank mRNA | AF319976.1 | - | 6 - 7 | 1085 | 64 | |
| AJ277114.1 | - | 7 - 8 | 1161 | 64 | ||
| AK138441.1 | - | 7 - 8 | 1134 | 64 | ||
| AK169006.1 | - | 7 - 8 | 1167 | 64 | ||
| BC023319.1 | - | 6 - 7 | 1337 | 64 | ||
| BC058581.1 | - | 7 - 8 | 1174 | 6491028 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|

Maged2
Genomic Map