Assay Details
| Gene Symbol: Erap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030711.4 | NP_109636.1 | 11 - 12 | 1720 | 92 | |
| GenBank mRNA | AB047552.1 | - | 10 - 11 | 1708 | 92 | |
| AF227511.1 | - | 10 - 11 | 1696 | 92 | ||
| AK029841.1 | - | 11 - 12 | 1719 | 92 | ||
| AK030329.1 | - | 11 - 12 | 1721 | 92 | ||
| AK036559.1 | - | 1721 | 92 | |||
| AK040350.1 | - | 8 - 9 | 938 | 92 | ||
| AK159406.1 | - | 11 - 12 | 1721 | 92 | ||
| BC046610.1 | - | 11 - 12 | 1726 | 6397585 | 92 | |
More Information
Updated Mapping Information
Set Membership: |
|

Erap1
Genomic Map