Assay Details
| Gene Symbol: Nckipsd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030729.4 | NP_109654.2 | 1 - 2 | 273 | 61 | |
| GenBank mRNA | AF130313.2 | - | 1 - 2 | 169 | 61 | |
| AK134725.1 | - | 1 - 2 | 255 | 61 | ||
| BC064818.1 | - | 1 - 2 | 273 | 30292108 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26816719 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26816722 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26816721 | - | - |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329541 | BC064818.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329541 | AK134725.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329541 | NM_030729.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329541 | AF130313.2 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329541 | BC064818.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329541 | AK134725.1 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329541 | NM_030729.4 | mouse_transcriptome |
| Agilent Whole Mouse Genome Microarray 4x44K v2 | A_51_P329541 | AF130313.2 | mouse_transcriptome |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 290208 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 492125 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 957696 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 215677 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 128075 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 35527 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 1x1M | 206445 | - | - |
| Agilent SurePrint G3 Mouse CGH Microarray 4x180K | 119755 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P133629 | - | - |
| Agilent Mouse Genome CGH Microarray 44K G4426B | A_53_P170146 | - | - |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329541 | BC064818.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329541 | AK134725.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329541 | NM_030729.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329541 | AF130313.2 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329541 | BC064818.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329541 | AK134725.1 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329541 | NM_030729.4 | mouse_transcriptome |
| SurePrint G3 Mouse Gene Expression Microarray G4852A | A_51_P329541 | AF130313.2 | mouse_transcriptome |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816712 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816718 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816729 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816709 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816715 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816726 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816707 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816734 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816737 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816739 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816727 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816725 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816722 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816723 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816732 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816738 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816711 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816721 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816714 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816717 | - | - |
| Agilent Mouse Promoter ChIP-on-Chip A) | A_68_P26816730 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P133629 | - | - |
| Agilent Mouse Genome CGH Microarray 244A (G4415A) | A_53_P170146 | - | - |
| Whole Mouse Genome G4122A | A_51_P329541 | BC064818.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329541 | AK134725.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329541 | NM_030729.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329541 | AF130313.2 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329541 | BC064818.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329541 | AK134725.1 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329541 | NM_030729.4 | mouse_transcriptome |
| Whole Mouse Genome G4122A | A_51_P329541 | AF130313.2 | mouse_transcriptome |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P117397 | - | - |
| Agilent Mouse Developmental Microarray 44K G2519A Option 002 | A_66_P106727 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P133629 | - | - |
| Agilent Mouse Genome CGH Microarray 105A (G4416A) | A_53_P170146 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P07810387 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08063639 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08063634 | - | - |
| Agilent Mouse Proximal Promoter ChIP-on-Chip A) | A_68_P08063640 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26816721 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26816719 | - | - |
| Agilent Mouse CpG Island ChIP-on-Chip Microarray 2x105K | A_68_P26816722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816763 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816840 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816709 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816781 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816715 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816757 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816775 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816714 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816734 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816809 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816796 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816749 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816737 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816668 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816792 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816850 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816777 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816735 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816801 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816670 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816836 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816722 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816771 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816849 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816766 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816664 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816851 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816770 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816750 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816772 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816838 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816791 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816742 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816751 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816707 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816834 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816844 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816729 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816846 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816811 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816800 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816761 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816756 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816831 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816828 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816725 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816853 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816826 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816857 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816807 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816787 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816665 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816804 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816774 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816711 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816732 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816718 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816676 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816748 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816780 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816824 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816835 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816830 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816790 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816816 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816768 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816746 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816855 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816753 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816821 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816723 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816795 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816842 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816755 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816674 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816822 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816788 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816726 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816730 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816778 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816783 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816717 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816744 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816833 | - | - |
| Agilent Mouse Whole Genome ChIP-on-Chip | A_68_P26816739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07879713 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816787 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816780 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816763 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816709 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816761 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P08063641 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816760 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07643544 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816753 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816771 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07556793 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07928397 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816790 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816739 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816717 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07928388 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816757 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07972764 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816732 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07928394 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07928383 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816788 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816715 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816744 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07556789 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816742 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07681301 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816774 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07810386 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816723 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816726 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816777 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07556794 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816768 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816772 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816755 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816785 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816730 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816778 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816795 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816738 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816766 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816770 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816775 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816707 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816714 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816722 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07680457 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07879716 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816746 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P07641482 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816735 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816791 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816718 | - | - |
| Agilent Six_ChIP_02_of_03_v2 | A_68_P26816792 | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Mouse Gene 1.0 ST Array | 10589125 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589117 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589119 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589121 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589116 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589114 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589115 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589120 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589122 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589124 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589123 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589126 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589127 | - | - |
| Affymetrix Mouse Gene 1.0 ST Array | 10589118 | - | - |
| Affymetrix HT MG-430 PM Array Plate | 1418975_PM_at | - | - |
| Affymetrix Mouse Gene 1.1 ST Array | 10589113 | - | - |
| Mouse Genome 430 2.0 Array | 1418975_PM_at | - | - |
| Mouse Genome 430 2.0 Array | 1418975_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 80987_at | - | - |
| Affymetrix GeneChip Mouse Genome 430 2.0 Array | 80987_PM_at | - | - |
| Affymetrix Mouse Clariom S Array | PSR0900011528.mm.1 | NM_030729.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011528.mm.1 | NM_030729.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011528.mm.1 | TC0900001423.mm.1 | MTA TC |
| Affymetrix Mouse Clariom S Array | PSR0900011529.mm.1 | NM_030729.4.1 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011529.mm.1 | NM_030729.4 | Refseq |
| Affymetrix Mouse Clariom S Array | PSR0900011529.mm.1 | TC0900001423.mm.1 | MTA TC |
| Mouse Expression Set 430 | 1418975_PM_at | - | - |
| Mouse Expression Set 430 | 1418975_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418975_at | - | - |
| Mouse Genome 430A 2.0 Array | 1418975_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418975_PM_at | - | - |
| Affymetrix HT Mouse Genome 430A Array | 1418975_at | - | - |
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| MouseWG-6 v2 Expression BeadChip | 2940156 | - | - |
| Illumina mouse-6 v1.0 expression beadchip | 2940156 | - | - |
| Illumina mouseRef-8 v1.0 expression beadchip | 0002940156 | - | - |
| Illumina mouseRef-8 v1.1 expression beadchip | ILMN_2619380 | - | - |
| Illumina mouse-6 v1.1 expression beadchip | ILMN_2619380 | - | - |
| mouse-6 v1.1 (Illumina) | scl36455.12.1_30-S | - | - |
| Illumina Sentrix mouse-6.v1 bead array | 2940156 | - | - |
| Illumina Sentrix MouseRef-8 Expression BeadChip | scl36455.12.1_30-S | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (2) | 2940156 | - | - |
| MouseRef-8 v2 Expression BeadChip | ILMN_2619380 | - | - |
| Illumina MouseWG-6 v2.0 expression beadchip | ILMN_2619380 | - | - |
| Illumina Sentrix Mouse-6 Expression BeadChip (3) | scl36455.12.1_30-S | - | - |

Nckipsd
Genomic Map