Assay Details
| Gene Symbol: Rad54l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030730.2 | NP_109655.2 | 2 - 3 | 428 | 61 | |
| GenBank mRNA | AJ132389.1 | - | 2 - 3 | 329 | 61 | |
| AK083903.1 | - | 2 - 3 | 409 | 61 | ||
| AK143506.1 | - | 1 - 2 | 44 | 61 | ||
| AK155151.1 | - | 2 - 3 | 385 | 61 | ||
| BC133714.1 | - | 2 - 3 | 277 | 40143565 | 61 | |
| CJ123928.1 | - | 2 - 3 | 62 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Rad54l2
Genomic Map