Assay Details
| Gene Symbol: Rad54l2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030730.2 | NP_109655.2 | 10 - 11 | 1975 | 82 | |
| GenBank mRNA | AJ132389.1 | - | 10 - 11 | 1876 | 82 | |
| AK083903.1 | - | 10 - 11 | 1956 | 82 | ||
| AK143506.1 | - | 9 - 10 | 1591 | 82 | ||
| AK155151.1 | - | 10 - 11 | 1932 | 82 | ||
| BC133714.1 | - | 10 - 11 | 1824 | 40143565 | 82 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rad54l2
Genomic Map