Assay Details
| Gene Symbol: Tbl1xr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030732.3 | NP_109657.2 | 2 - 3 | 369 | 69 | |
| GenBank mRNA | AF268195.1 | - | 2 - 3 | 150 | 69 | |
| AK029595.1 | - | 2 - 3 | 168 | 69 | ||
| AK033347.1 | - | 2 - 3 | 239 | 69 | ||
| AK036064.1 | - | 3 - 4 | 480 | 69 | ||
| AK136255.1 | - | 2 - 3 | 191 | 69 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AF268195.1,AK029595.1,AK033347.1,AK036064.1,AK136255.1,NM_030732.3 transcripts.
Set Membership: |
|

Tbl1xr1
Genomic Map