| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0300000705.mm.1 |
NM_030732.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000705.mm.1 |
NM_030732.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000705.mm.1 |
TC0300000144.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300000704.mm.1 |
NM_030732.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000704.mm.1 |
NM_030732.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000704.mm.1 |
TC0300000144.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0300000706.mm.1 |
NM_030732.3.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000706.mm.1 |
NM_030732.3 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0300000706.mm.1 |
TC0300000144.mm.1 |
MTA TC |