Assay Details
| Gene Symbol: Tbl1xr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030732.3 | NP_109657.2 | 8 - 9 | 1180 | 129 | |
| GenBank mRNA | AF268195.1 | - | 8 - 9 | 961 | 129 | |
| AK029595.1 | - | 8 - 9 | 979 | 129 | ||
| AK033347.1 | - | 8 - 9 | 1050 | 129 | ||
| AK036064.1 | - | 9 - 10 | 1291 | 129 | ||
| AK136255.1 | - | 8 - 9 | 1002 | 129 | ||
More Information
Set Membership: |
|

Tbl1xr1
Genomic Map