Assay Details
| Gene Symbol: Tbl1xr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030732.3 | NP_109657.2 | 13 - 14 | 1666 | 78 | |
| GenBank mRNA | AF268195.1 | - | 13 - 14 | 1447 | 78 | |
| AK029595.1 | - | 13 - 14 | 1465 | 78 | ||
| AK033347.1 | - | 13 - 14 | 1536 | 78 | ||
| AK036064.1 | - | 14 - 15 | 1777 | 78 | ||
| AK136255.1 | - | 13 - 14 | 1488 | 78 | ||
| BC018512.1 | - | 3 - 4 | 312 | 78 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Tbl1xr1
Genomic Map