Assay Details
| Gene Symbol: Sil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030749.2 | NP_109674.2 | 1 - 2 | 131 | 85 | |
| GenBank mRNA | AJ297884.1 | - | 1 - 2 | 72 | 85 | |
| AK170415.1 | - | 1 - 2 | 84 | 85 | ||
| BC016119.1 | - | 1 - 2 | 108 | 4489217 | 85 | |
| BC016466.1 | - | 1 - 2 | 94 | 3582798 | 85 | |
| BC028488.1 | - | 1 - 2 | 68 | 85 | ||
| CX219879.1 | - | 1 - 2 | 131 | 85 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AJ297884.1,AK170415.1,BC016119.1,BC016466.1,NM_030749.2 transcripts.
Set Membership: |
|

Sil1
Genomic Map