Assay Details
| Gene Symbol: Sil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030749.2 | NP_109674.2 | 6 - 7 | 806 | 79 | |
| GenBank mRNA | AJ297884.1 | - | 6 - 7 | 747 | 79 | |
| AK170415.1 | - | 6 - 7 | 759 | 79 | ||
| AK192274.1 | - | 1 - 2 | 126 | 79 | ||
| AK219586.1 | - | 1 - 2 | 126 | 79 | ||
| BC016119.1 | - | 6 - 7 | 783 | 4489217 | 79 | |
| BC016466.1 | - | 6 - 7 | 769 | 3582798 | 79 | |
| CO428760.1 | - | 3 - 4 | 349 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sil1
Genomic Map