Assay Details
| Gene Symbol: Sil1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_030749.2 | NP_109674.2 | 7 - 8 | 919 | 79 | |
| GenBank mRNA | AJ297884.1 | - | 7 - 8 | 860 | 79 | |
| AK170415.1 | - | 7 - 8 | 872 | 79 | ||
| AK219586.1 | - | 2 - 3 | 239 | 79 | ||
| BC016119.1 | - | 7 - 8 | 896 | 4489217 | 79 | |
| BC016466.1 | - | 7 - 8 | 882 | 3582798 | 79 | |
| CO428760.1 | - | 4 - 5 | 462 | 79 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Sil1
Genomic Map