Assay Details
| Gene Symbol: Sf3b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_031179.2 | NP_112456.2 | 1 - 2 | 135 | 70 | |
| GenBank mRNA | AB037890.1 | - | 1 - 2 | 160 | 70 | |
| AK012632.1 | - | 1 - 2 | 94 | 70 | ||
| AK140982.1 | - | 1 - 2 | 53 | 70 | ||
| AK146989.1 | - | 1 - 2 | 44 | 70 | ||
| AK165992.1 | - | 1 - 2 | 139 | 70 | ||
| AK171454.1 | - | 1 - 2 | 53 | 70 | ||
| BC003419.1 | - | 1 - 2 | 38 | 70 | ||
| BC028959.1 | - | 1 - 2 | 38 | 70 | ||
| BC037098.1 | - | 1 - 2 | 122 | 70 | ||
| BC058563.1 | - | 1 - 2 | 38 | 70 | ||
| BC080754.1 | - | 1 - 2 | 54 | 70 | ||
| CK331410.1 | - | 1 - 2 | 135 | 70 | ||
More Information
Updated Mapping Information
Additional Information:
This assay is in the 5'UTR region of the AB037890.1,AK012632.1,AK140982.1,AK171454.1,BC003419.1,BC028959.1,NM_031179.2 transcripts.
Set Membership: |
|

Sf3b1
Genomic Map